n-Dodecyl-β-D-maltoside ≥99% (by HPLC), high purity

Supplier: Enzo Life Sciences

500-007-G001 500-007-G005 500-007-G025
89151-634EA 251.22 CAD
89151-634 89151-636 89151-638
n-Dodecyl-β-D-maltoside ≥99% (by HPLC), high purity
n-Dodecyl-β-D-maltoside

Non-ionic detergent free of alpha-isomer (HPLC)


  • Formulation: White crystal powder.


Non-ionic detergent similar to n-octyl-beta-D-glucopyranoside (Prod. No. ALX-500-001). However, increased alkyl chain length has been shown to increase thermal stability of lipid-free rhodopsin in solution. Applicable to membrane-protein solubilization studies.

Formula: C₂₄H₄₆O₁₁
MDL Number: MFCD00043012
CAS Number: 69227-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1530df19ea3b491440f901498f6c98c2dbd1ab66&vid=b8296acafb221a9c76daa6151416c31145678613&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR