Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥98%

Supplier: Enzo Life Sciences

Synonyms: Hecameg®

500-003-G001 500-003-G005
89151-630EA 105.86 CAD
89151-630 89151-632
Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside ≥98%
Methyl-6-O-(N-heptylcarbamoyl)-alpha-D-glucopyranoside

Non-ionic, glucose based very mild detergent for membrane protein purification.


  • Formulation: White to off-white crytalline solid.


Hecameg is used for the extraction, purification, stabilization of proteins (recombinant or natural proteins). Surfactant for chromatography, electrophoresis and ELISA analysis. Extraction of other biomolecules (DNA and RNA) from proteous samples. Also used for the study of protein structure; reconstitution or crystallization of membrane proteins, enzymes or antigens; liposomes preparation. Sanitization of chromatography columns.


Soluble in water (>100 mg/mL at +4 °C).


Synonym: 6-O-(N-Heptylcarbamoyl)methyl-α-D-glucopyranoside

Formula: C₁₅H₂₉NO₇
MW: 335.397 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077425
CAS Number: 115457-83-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24ff637b6676a078c5b9a139c5f4fe92a74623b0&vid=f743675fc1b930da6c8526cc6b9bf600a51d503d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Appearance White to off-white crytalline solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR