N-Octyl-β-D-glucopyranoside ≥99% (by HPLC) analytical reagent

Supplier: Enzo Life Sciences

500-001-G001 500-001-G005 500-001-G025
89151-618EA 109.02 CAD
89151-618 89151-620 89151-622
N-Octyl-β-D-glucopyranoside ≥99% (by HPLC) analytical reagent
N-Octyl-β-D-glucopyranoside

Non-ionic detergent. Highly purified.


  • Formulation: White to off-white powder.


Highly purified nonionic detergent that solubilizes membrane-bound proteins in their native state.

Formula: C₁₄H₂₈O₆
MDL Number: MFCD00063288
CAS Number: 29836-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d52f048eb7995ee20203563e8b2710c80ac5b4a&vid=23f2980244f88e16f16b32e79f2e923e622157d0&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR