Gemcitabine hydrochloride ≥98%

Supplier: Enzo Life Sciences

Synonyms: 2'-Deoxy-2',2'-difluorocytidine, dFdC, 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

480-101-M025 480-101-M100
89151-614EA 184.86 CAD
89151-614 89151-616
Gemcitabine hydrochloride ≥98%
Gemcitabine hydrochloride

DNA synthesis inhibitor


Deoxycytidine analog with cytotoxic effects. Induces apoptosis and inhibits DNA synthesis.


Synthetic.

Formula: C₉H₁₂ClF₂N₃O₄
MDL Number: MFCD01735988
CAS Number: 122111-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f136f573c46b971b427de0803d3e9c4d3ec3d41e&vid=eb9c70577f66ba9dc41a70b3e78021e4bd0f693d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR