Arabinofuranosyl-2-fluoroadenine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine

480-100-M005 480-100-M025
89151-610EA 105.86 CAD
89151-610 89151-612
Arabinofuranosyl-2-fluoroadenine ≥98% (by HPLC)
Arabinofuranosyl-2-fluoroadenine

F-ara-A precursor


Prodrug converted to the free nucleoside 9-beta-D-arabinosyl-2-fluoroadenine (F-ara-A). F-ara-A enters cells and accumulates as the 5'-triphosphate. Interferes with DNA synthesis and repair. Induces apoptosis of cancer cells. Inhibits RNA transcription.

Formula: C₁₀H₁₂FN₅O₄
MW: 285.24 g/mol
MDL Number: MFCD00132942
CAS Number: 21679-14-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bb0099c44d58405c13977f3ff798beb90f2d9df&vid=1956b28452730714a0f1efde75f06235f0c72dac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR