8-(4-Chlorophenylthio)-2'-O-methyladenosine-3’,5’-cyclic monophosphate sodium salt ≥98% (by HPLC)

Supplier: Enzo Life Sciences

480-093-M001 480-093-M005
89151-588EA 402.9 CAD
89151-588 89151-590
8-(4-Chlorophenylthio)-2'-O-methyladenosine-3’,5’-cyclic monophosphate sodium salt ≥98% (by HPLC)
8-(4-Chlorophenylthio)-2'-O-methyladenosine-3’,5’-cyclic monophosphate sodium salt

Specific, potent and membrane permeant activator of the Epac cAMP receptor.


  • Formulation: White to off-white crystalline powder.


Activates Epac1 and Epac2, the guanine nucleotide-exchange factors (GEFs) for the small GTPases Rap1 and Rap2. Does not activate protein kinase A (PKA), neither in vitro nor in vivo and is therefore a valuable tool to specifically discriminate between PKA- and Epac-mediated effects. Since a free 2'-ribose hydroxyl group in cyclic AMP is essential for stimulation of PKA, the methylated structure of 8-CPT-2'-O-Me-cAMP is an extremely poor PKA activator.


Its high lipophilicity allows for good membrane permeability in most biosystems and its increased resistance towards phosphodiesterases prevents rapid hydrolysis.


Hygroscopic and soluble in water, aqueous buffers, DMSO, dimethyl formamide or methanol.


Synonym: 8-CPT-2'-O-Me-cAMP.Na

Formula: C₁₇H₁₆ClN₅O₆PS·Na
MW: 507.8 g/mol
Storage Temperature: Freezer
CAS Number: 510774-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a77a4c223faf5948d704e6d9cac92d65a637871&vid=c5f6fe0c6e94893fb87b2fd0ca2dfe59be9ef3e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Identity Determined by ESI-MS and UV
Purity detail Product is not sterile

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR