8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (by HPLC)

Supplier: Enzo Life Sciences

480-088-M001 480-088-M005
89151-580EA 79 CAD
89151-580 89151-582
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (by HPLC)
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt

Selective activator of cGMP-dependent protein kinase (PKG) Iα, Iβ, and type II as well as of cGMP-gated ion channels.


  • Formulation: White to off-white powder.


Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP.


Soluble in water.


Synonym: 8-pCPT-cGMP.Na

Formula: C₁₆H₁₄ClN₅O₇PS·Na
Storage Temperature: Freezer
CAS Number: 51239-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f9e095fcbb33c246288d1b2c574b28fdbbf88ab&vid=15fd3a286830e3f6dc18117e3bb2033a075dac3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥99%
Identity Determined by ESI-MS and UV/VIS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR