Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85%
Supplier: Enzo Life Sciences
|
Warning
|
480-056-M001
480-056-M005
89151-530EA
110.6
CAD
89151-530
89151-532
Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt ≥85%
Guanosine-5'-O-(3-thiotriphosphate) tetralithium salt
Nonhydrolyzable GTP analog.
- Formulation: White to off-white solid.
A nonhydrolysable analog of guanosine-5’-O-triphosphate (GTP). Stimulates inositol trisphosphate formation in membranes.
Soluble in water (25 mg/mL).
Synonym: GTP-γ-S.4Li
Formula:
C₁₀H₁₂N₅O₁₃P₃S·4Li Storage Temperature: Freezer |
CAS Number:
94825-44-2 |
|
Specification Test Results
Purity (HPLC) | ≥85% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...