8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt ≥99% (by HPLC)

Supplier: Enzo Life Sciences

480-032-C100 480-032-M001
89151-520EA 124.82 CAD
89151-520 89151-522
8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt ≥99% (by HPLC)
8-(4-Chlorophenylthio)guanosine 3',5'-cyclic Monophosphothioate, Rp-Isomer triethylammonium salt

Potent selective inhibitor of cGMP-dependent protein kinase 1α, 1β and type II. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases, no metabolic side effects. Significantly more lipophilic and membrane permeant as compared to Rp-cGMPS or Rp-8-Br-cGMPS.


  • PKG inhibitor


White to off-white crystalline powder.


Soluble in water and aqueous buffers.

Formula: C₁₆H₁₄ClN₅O₆PS₂·C₆H₁₆N
Storage Temperature: Freezer
CAS Number: 153660-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07bb6bab89f53c5d9ff804f48331225b801e1973&vid=4172a140899e77ce74b7cc46c1e7ad8f1c626163&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR