8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Supplier: Enzo Life Sciences
480-024-M001
480-024-MC05
89151-500EA
527.72
CAD
89151-500
89151-502
8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt
Inhibitor of cAMP-dependent protein kinase (PKA), preferentially binding to type I PKA. Lipophilic analog of Rp-cAMPS. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases. Significantly more lipophilic and membrane permeant as compared to Rp-cAMPS or 8-Br-cAMP.
- PKA inhibitor
White to off-white powder.
250 mM soluble in water, aqueous solutions, DMSO or dimethyl formamide.
Formula:
C₁₀H₁₀BrN₅O₅PS·Na Storage Temperature: Freezer |
CAS Number:
129735-00-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...