ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) ≥97% (by HPLC)

Supplier: Enzo Life Sciences

480-003-G001 480-003-M050 480-003-M250
89151-468EA 417.12 CAD
89151-468 89151-470 89151-472
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) ≥97% (by HPLC)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)


  • Formulation: White to light yellow powder.

Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MDL Number: MFCD00065390
CAS Number: 24292-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb07b0ad1f0cd505131b3d56824dfcb8284420ed&vid=2f25b621a9968349af8c4d94a986a7eb42e27e32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR