Adenosine-5'-diphosphate monopotassium salt ≥87%
Supplier: Enzo Life Sciences
Synonyms:
ADP.K, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate monopotassium salt, 5'-ADP-K
480-002-G001
480-002-G005
89151-464EA
109.02
CAD
89151-464
89151-466
Adenosine-5'-diphosphate monopotassium salt ≥87%
Adenosine-5'-diphosphate monopotassium salt
- Formulation: White to off-white crystalline powder.
Isolated from yeast.
Soluble in water (50 mg/mL).
Synonym: ADP.K
Formula:
C₁₀H₁₄N₅O₁₀P₂K MW: 465.29 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00066472 CAS Number: 70285-70-0 Merck Index: 14,00154 |
Specification Test Results
Purity (assay) | ≥87% |
Source | Isolated from yeast |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...