Adenosine 5'-diphosphate disodium salt (ADP disodium salt)

Supplier: Enzo Life Sciences

Synonyms: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate

480-001-G001 480-001-G005
89151-460EA 120.08 CAD
89151-460 89151-462
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)

Isolated from yeast.

Formula: C₁₀H₁₃N₅Na₂O₁₀P₂
MDL Number: MFCD00066635
CAS Number: 16178-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9019cf0d62238d041d7efa984e07f5dd5053566f&vid=799c5b287d55ad659af0d6eabab629a942839abe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR