Nonactin (from Streptomyces griseus) ≥98% (by TLC)

Supplier: Enzo Life Sciences

450-008-M005 450-008-M025
89151-390EA 91.64 CAD
89151-390 89151-392
Nonactin (from Streptomyces griseus) ≥98% (by TLC)
Nonactin

Macrotetrolide antibiotic. Monovalent cation ionophore with high selectivity for ammonium and potassium. Specifically inhibits the processing of cytoplasmic precursor proteins destined for the mitochondria. Inhibits the effects on P170 glycoprotein-mediated efflux of chemotherapeutic agents in multidrug resistant (MDR) cancer cells.


  • Monovalent cation ionophore


White to off-white crystalline solid.


Soluble in DMSO (10mg/ml), dichloromethane (10mg/ml), methanol, 100% ethanol; insoluble in water.

Formula: C₄₀H₆₄O₁₂
Storage Temperature: Freezer
CAS Number: 6833-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a778a1d33c64ca8a8f254fad8362781e8f3474&vid=70b6d05d0d7fea300163f8a53f2fc04e79c6f45a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR