Ionomycin calcium salt (from Streptomyces conglobatus) ≥97% (by HPLC, TLC)

Supplier: Enzo Life Sciences

450-007-M001 450-007-M005
89151-386EA 237 CAD
89151-386 89151-388
Ionomycin calcium salt (from Streptomyces conglobatus) ≥97% (by HPLC, TLC)
Ionomycin calcium salt

Potent and highly selective calcium ionophore


Ca2+/ionomycin complex forms between pH 7-9.5 which has intense UV absorption. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase. Useful tool for increasing intracellular Ca2+ concentration. More effective than A23187. Non fluorescent. Has been used at 4 nM and 2-4 µM  (whole cells).


White to off-white solid


Solubility: 10 mg/mL soluble in DMSO or 100% ethanol, 5 mg/mL soluble in methanol


Packaging: Shipping temperature: Ambient

Formula: C₄₁H₇₀CaO₉
Storage Temperature: Refrigerator
MDL Number: MFCD00151183
CAS Number: 56092-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47790c9a66a69950e15e47f7427094c5100aea7&vid=92a162e64703c3b9d8adfc26d545b2b1bc1d7da8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR