Ionomycin free acid (from Streptomyces conglobatus) ≥90% (by HPLC, TLC)

Supplier: Enzo Life Sciences

450-006-M001 450-006-M005
89151-382EA 184.86 CAD
89151-382 89151-384
Ionomycin free acid (from Streptomyces conglobatus) ≥90% (by HPLC, TLC)
Ionomycin

Potent and highly selective calcium ionophore


Ca2+/ionomycin complex forms between pH 7-9.5 which has intense UV absorption. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest at G1 phase. Useful tool for increasing intracellular Ca2+ concentration. More effective than A23187. Non fluorescent. Has been used at 4 nM and 2-4 µM  (whole cells).


Waxy yellow to colorless material


Soluble in DMSO or methanol


Packaging: Shipping temperature: Ambient

Formula: C₄₁H₇₂O₉
MW: 709.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06798385
CAS Number: 56092-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61e7f78e2db595a0d7d817052e90c2be00ad623b&vid=35c6aeaaa543fd49113a7f3e7141e0b0f872b275&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR