Necrostatin-5 ≥98% (by NMR)

Supplier: Enzo Life Sciences

430-169-M005 430-169-M025
89151-304EA 96.38 CAD
89151-304 89151-306
Necrostatin-5 ≥98% (by NMR)
Necrostatin-5

Necroptosis inhibitor


Necroptosis inhibitor. In contrary to the mechanism of necrostatin-1 (Prod.No. BML-AP309) it was shown that necrostatin-5 inhibits RIP1 kinase indirectly. A potent inhibitor of immunoprecipitated, but not recombinant RIP1.

Formula: C₁₉H₁₇N₃O₂S₂
CAS Number: 337349-54-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3b68ff57081621e553440ab99f0d5d79ba9b543&vid=2a6cc39b2bda6a2b2cbd427d19996ebff19ef13b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR