Necrox-5™ methanesulfonate

Supplier: Enzo Life Sciences

Synonyms: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine

430-167-M001 430-167-M005
89151-300EA 251.22 CAD
89151-300 89151-302
Necrox-5™ methanesulfonate
Necrox-5™ methanesulfonate

Necrosis inhibitor.


Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models.

Formula: C₂₅H₃₁N₃O₃S·2CH₄O₃S
MW: 645.8 g/mol
CAS Number: 1383718-29-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17941399b617f6cb3fc0e1e9ace76794f4318037&vid=06a96f3249c03864d9b41f1627d3dfe17b164430&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR