Lamotrigine ≥98%

Supplier: Enzo Life Sciences

Synonyms: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

430-160-M025 430-160-M100 430-160-M500
89151-278EA 346.02 CAD
89151-278 89151-280 89151-282
Lamotrigine ≥98%
Lamotrigine

Na+ channel blocker


Lamotrigine blocks voltage-gated sodium channels. Inhibits the release of glutamate. Neuroprotective. Anticonvulsant.

Formula: C₉H₇Cl₂N₅
MW: 256.09 g/mol
MDL Number: MFCD00865333
CAS Number: 84057-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c13b379abc5b0812c1b24fb6873fe08be397b957&vid=9497ed10575b0084805ae8391c22e7f73cec6d26&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR