Zoledronic acid ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid, ZOL-446, CGP 4244, CGP 42446A, Zoledronate

430-153-G001 430-153-M025 430-153-M100
89151-260EA 1275.06 CAD
89151-260 89151-262 89151-264
Zoledronic acid ≥98% (by HPLC)
Zoledronic acid

Acid sphingomyelinase inhibitor


Inhibitor of osteoclastic bone resorption. Prevents osteoporosis and skeletal fractures, particularly in patients with multiple myeloma or prostate cancer. Also used to treat hypercalcemia.Inhibition of acid sphingomyelinase (aSMase) with an IC50 5µm.


Synthetic.

Formula: C₅H₁₀N₂O₇P₂
MW: 272.09 g/mol
MDL Number: MFCD00867791
CAS Number: 118072-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=916013288885c3927939d2ec7f5254599a599178&vid=bd49bdc97860ef6409051ad91809db4f11056fe1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR