Procaspase-3-activator (PAC-1) ≥98% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences
Warning

430-146-M010 430-146-M050
89151-238EA 105.86 CAD
89151-238 89151-240
Procaspase-3-activator (PAC-1) ≥98% (by HPLC), white to off-white solid
Procaspase-3 Activator (PAC1, (4-Benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide)

Caspase-3 activator.


  • Formulation: White to off-white solid.


First molecule to directly activate procaspase-3 to caspase-3 in vitro. Has been shown to induce apoptosis in cancer cells.


Soluble in acetonitrile, DMSO, dioxane or methanol.

Formula: C₂₃H₂₈N₄O₂
MDL Number: MFCD03302441
CAS Number: 315183-21-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1caa0e1425829a2e1101c35c8b34683169622664&vid=942ab647585fd48fb98e59d5b0bfb65deeb8d4ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC (UV)) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR