IM-54 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1-Methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-1H-pyrrole-2,5-dione

430-137-M001 430-137-M005
89151-204EA 71.1 CAD
89151-204 89151-206 89151-208
IM-54 ≥98% (by HPLC)
IM-54

Cell permeable inhibitor of necrosis


Selectively blocks oxidative stress-induced necrotic cell death (~3 µm IM-54 prevented ~50% cell death in HL60 cells exposed to 100 µm H2O2). Does not protect against etoposide-induced apoptosis or display antioxidant properties. Does not affect S6K1 and PKC kinase activities at concentrations of 50 µm.

Formula: C₁₉H₂₃N₃O₂
MDL Number: MFCD18382109
CAS Number: 861891-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c407f0979ebbc2f5b815ec914ee2b7964567597d&vid=b703195e54ce56c65be725984e3c06dfbf80508a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR