TMH hydrochloride
Supplier: Enzo Life Sciences
Synonyms:
1-Hydroxy-4-methoxy-2,2,6,6-tetramethylpiperidine·HCl
430-132-M050
430-132-M250
89151-186EA
453.46
CAD
89151-186
89151-188
TMH hydrochloride
TMH hydrochloride
Cyclic hydroxylamine spin probe for in vivo, intraperitoneal and intravenous injections.
- Formulation: White to off-white solid.
Lifetime of more than 4 hours in blood plasma, in cells and tissue. Allows quantitative measurements of extracellular O²⁻ and quantification of intracellular O²⁻ in cells and tissue samples and in vivo O²⁻ detection.
Soluble in water, methanol or 100% ethanol.
|
Specification Test Results
Purity detail (EPR) | Paramagnetic impurities ≤0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...