Nutlin-3 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 4-(cis-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one, (±)-4-[4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-once

430-128-M001 430-128-M005 430-128-M025
89151-168EA 88.48 CAD
89151-168 89151-170 89151-172
Nutlin-3 ≥98% (by HPLC)
Nutlin-3

Inhibitor of p53/MDM2 interaction


(+/-) -Nutlin-3 is an inhibitor of p53/MDM2 interaction (IC50=0.09µM).

Formula: C₃₀H₃₀Cl₂N₄O₄
MDL Number: MFCD07784509
CAS Number: 548472-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b8f2aa80d6cc920f9e92b716dbaca24c3a1305d&vid=b4ccbccd39e9f5ecb5e47e6793e1c7ddeff46e8c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR