CMH hydrochloride ≥99%
Supplier: Enzo Life Sciences
430-117-M010
430-117-M050
430-117-M250
89151-138EA
162.74
CAD
89151-138
89151-140
89151-142
CMH hydrochloride ≥99%
CMH hydrochloride
Spin trapping reagent.
- Formulation: White to light-yellow crystalline powder.
Analog of CPH with higher cell permeability and 3-fold better reactivity with superoxide.
It has been used to study intracellular production of superoxide radicals in plasma, endothelial cells and isolated hearts.
Soluble in DMSO, 100% ethanol, methanol or water.
Caution: Prepare fresh solutions daily before use and store 4-6 hours on ice under a nitrogen/argon flow.
|
Specification Test Results
Purity (EPR) | ≥99% |
Formulation | White to light-yellow crystalline powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...