RSSR ≥97% (by HPLC), crystalline powder
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
bis-(2,2,5,5-Tetramethyl-3-imidazoline-1-oxyl-4-yl)disulfide, biradical, bis-(2,2,5,5-Tetramethyl-3-imidazoline-1-oxyl-4-yl)disulphide, biradical
Supersensitive spin label.
- Formulation: Off-white to yellow crystalline powder.
Cell permeable supersensitive spin label (detection limit varies from 10nM to 100nM) for quick detection of reduced thiols using ESR. Allows following the reactions of sulfhydryl groups with RSSR to form thiol spin label adducts, for the monitoring of intracellular redox states of glutathione and other thiols.
Soluble in DMSO, water, 100% ethanol or methanol; DMSO is recommended for stock solution.
Synonyms: bis-(2,2,5,5-Tetramethyl-3-imidazoline-1-oxyl-4-yl)disulfide; biradical
Formula:
C₁₄H₂₄N₄O₂S₂ MW: 344.5 g/mol Storage Temperature: Freezer |
CAS Number:
123557-49-3 |
Specification Test Results
Purity (HPLC, GLC) | ≥97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...