Leflunomide ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5-Methyl-N-(4-(trifluoromethyl)phenyl)isoxazole-4-carboxamide, 5-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide

430-095-G001 430-095-M050
89151-092EA 982.76 CAD
89151-092 89151-096
Leflunomide ≥99% (by HPLC)
Leflunomide

Immunosuppressant


Immunosuppressive drug that inhibits T and B cell responses in vivo. Inhibitor of tyrosine phosphorylation (tyrosine kinase inhibitor), of dihydroorotate dehydrogenase, an enzyme for de novo pyrimidine synthesis, of TNF-dependent NF-kappaB activation, and of COX-2. The physiological activity of leflunomide is attributed mainly to its metabolite A77 1726 (Prod. No. ALX-430-096).

Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
MDL Number: MFCD00867593
CAS Number: 75706-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612a1f7eb2f90a49557e2e27d9330ac307cb3af7&vid=83c2ecce41185f4654cb5ed67324dcf086e2f11b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR