5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide ≥99%

Supplier: Enzo Life Sciences

Synonyms: DEPMPO

430-093-M050 430-093-M500
89151-088EA 364.98 CAD
89151-088 89151-090
5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide ≥99%
5-(Diethylphosphono)-5-methyl-1-pyrroline-N-oxide

Spin trapping reagent.


  • Formulation: Colorless to light yellow oil.


Most efficient spin trap for the in vitro and in vivo detection of O-, N-, S- and C-centered free radicals. Has a longer life-time than DMPO. Can distinguish between superoxide-dependent and independent mechanisms that lead to the hydroxyl radical. Less lipophilic (Kp=0.16) than DIPPMPO.


Soluble in water, methylene chloride or 100% ethanol.

Formula: C₉H₁₈NO₄P
MW: 235.22 g/mol
Storage Temperature: Freezer
CAS Number: 157230-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20ca7c49e3f33efc92c46b996176ca1dd732634a&vid=7f9e4cfcb20829599433c957538c4d43b5a05b80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (TLC) ≥99%
Appearance Colorless to light yellow oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR