5,5-Dimethyl-1-pyrroline-N-oxide ≥98% (by ELSD)

Supplier: Enzo Life Sciences

Synonyms: DMPO, 5,5-Dimethyl-1-pyrroline N-oxide

430-090-G001 430-090-M500
89151-078EA 504.02 CAD
89151-078 89151-080
5,5-Dimethyl-1-pyrroline-N-oxide ≥98% (by ELSD)
5,5-Dimethyl-1-pyrroline-N-oxide

Spin trapping reagent.


  • Formulation: Pale yellow solid or oil.


Cell permeable hydrophilic spin trap for both in vivo and in vitro studies of superoxide, O-, C-, S- and N- centered free radicals.


Soluble in water, 100% ethanol, methanol or DMSO.

Formula: C₆H₁₁NO
Storage Temperature: Freezer
MDL Number: MFCD00005279
CAS Number: 3317-61-1
EINECS: 222-011-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec4391380d73f490389a0b3e510016e0ee014bb1&vid=61714936834d4d3dee2f186988baa491f5ee7a04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (ELSD) ≥98%
Appearance Pale yellow solid or oil

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR