CPH hydrochloride ≥99% (EPR), white to off-white solid

Supplier: Enzo Life Sciences

Synonyms: 1-Hydroxy-3-methoxycarbonyl-2,2,5,5-tetramethylpyrrolidine . HCl

430-078-M010 430-078-M050 430-078-M250
89151-026EA 390.26 CAD
89151-026 89151-028 89151-030
CPH hydrochloride ≥99% (EPR), white to off-white solid
CPH hydrochloride

Spin trapping reagent.


  • Effective, cell permeable and non-toxic spin probe for the detection of superoxide radical and peroxynitrite, both in vitro and in vivo


Resistant to reduction by vitamin C and thiols.


Soluble in DMSO, 100% ethanol, methanol or water.

Formula: C₉H₁₇NO₃·HCl
MW: 223.7 g/mol
Storage Temperature: Freezer
CAS Number: 92416-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b50b9d84320c1b10d14459844316337e47842a76&vid=34a3fdd32cd28ab32b28390f4a94a6c651437473&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (EPR) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR