TEMPONE-H hydrochloride ≥99% (by TLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: 1-Hydroxy-2,2,6,6-tetramethyl-4-oxo-piperidine . HCl

430-071-M010 430-071-M050 430-071-M250
89151-006EA 146.94 CAD
89151-006 89151-008 89151-010
TEMPONE-H hydrochloride ≥99% (by TLC)
TEMPONE-H hydrochloride

Spin trapping reagent.


  • TEMPONE-H hydrochloride is a very effective, cell permeable and non-toxic spin trap for the detection of superoxide radical and peroxynitrite.


Soluble in DMSO, water, methanol or 100% ethanol.

Formula: C₉H₁₇NO₂·HCl
Storage Temperature: Freezer
CAS Number: 3637-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b7b7f408c9ee2dbfe2d6c66468ddac73d371d5c&vid=e58716d57dcf79bc739a986f73f1b47760745780&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (TLC) ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR