MnTMPyP pentachloride ≥95% (by TLC), black powder

Supplier: Enzo Life Sciences

430-070-M010 430-070-M050
89151-002EA 71.1 CAD
89151-002 89151-004
MnTMPyP pentachloride ≥95% (by TLC), black powder
MnTMPyP pentachloride

SOD mimetic. Peroxynitrite scavenger.


  • Formulation: Black crystalline powder.


Cell permeable superoxide dismutase (SOD) mimetic and peroxynitrite scavenger. Catalyzes the dismutation of O²⁻ even in the presence of excess EDTA. Displays a protective effect against H₂O₂ mediated injury in astroglial cells.


Soluble in water or aqueous buffers.

Formula: C₄₄H₃₆MnN₈Cl₅
Storage Temperature: Freezer
CAS Number: 100012-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56dc94b58db43294e335800b442ba0942abe0f9d&vid=455618ca2b3d1e2bc0bd7ae85c5fa0b2c553b0b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (TLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR