MnTBAP chloride ≥97%

Supplier: Enzo Life Sciences

430-069-M010 430-069-M050
89150-998EA 72.68 CAD
89150-998 89151-000
MnTBAP chloride ≥97%
MnTBAP chloride

SOD mimetic


Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Inhibits neuronal apoptosis.


Peroxynitrite scavenger.

Formula: C₄₈H₂₈MnN₄O₈Cl
MDL Number: MFCD06227760
CAS Number: 55266-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=872a25aa15e408a729337a84adb78ce1bc6d1536&vid=35aee9c6483efb24536fd07d5a90d5fa4d6b4375&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR