Tin Protoporphyrin IX dichloride ≥95% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: TIn protoporphyrin ix dichloride

430-051-M005 430-051-M025
89150-980EA 112.18 CAD
89150-980 89150-982
Tin Protoporphyrin IX dichloride ≥95% (by TLC)
Tin Protoporphyrin IX dichloride

Potent inhibitor of heme oxygenase.


  • Heme oxygenase inhibitor


Dark red to violet solid.


Soluble in aqueous base.

Formula: C₃₄H₃₂N₄O₄Sn·2Cl
MW: 750.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00216794
CAS Number: 14325-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92d9ec95703483c593dbc1d0de263cf1980a98a9&vid=9442b1656425c1d4ea5b126b35b76c1f45973d6d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR