Zinc protoporphyrin ≥95% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX

430-049-M025 430-049-M100
89150-968EA 74.31 CAD
89150-968 89150-970 89150-972
Zinc protoporphyrin ≥95% (by TLC)
Zinc protoporphyrin

Potent and selective inhibitor of heme oxygenase (Ki=3nM), the enzyme that generates carbon monoxide (CO). Inhibits soluble guanylyl cyclase. Produces a time- and concentration-dependent inactivation of all three isoforms of nitric oxide synthase (IC50=0.8µM, 4.0µM, and 5.0µM for nNOS (NOS I), iNOS (NOS II) and eNOS (NOS III), respectively). Does not cross the blood-brain barrier. An useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.


  • Heme oxygenase inhibitor


Dark solid.


Soluble in pyridine, dimethyl formamide or DMSO. Colloidal in aqueous base;  slightly soluble in aqueous base/organic solvent mixtures.

Formula: C₃₄H₃₂N₄O₄Zn
MW: 626.034 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00011612
CAS Number: 15442-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5e06b6be9bb405636fa02ee2f73fcf789eb9267&vid=a64e2d6169fd6da126b2390b1ca3c62a5eeaefb2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR