Protoporphyrin IX ≥95% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Ooporphyrin, Kammerer’s porphyrin, 3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid

430-041-G001 430-041-M300
89150-954EA 406.06 CAD
89150-954 89150-956
Protoporphyrin IX ≥95% (by HPLC)
Protoporphyrin IX

Protoporphyrin IX free acid, as distinct from its zinc salt (which inhibits heme oxygenase), activates soluble guanylyl cyclase (sGC) by binding directly to the enzyme. A useful reagent in cases where the use of nitric oxide (NO) or nitric oxide donors is undesirable.


  • Guanylyl cyclase activator


Purple to dark brown solid.


Dissolve initially in 0.1M base (Tris base or NaOH), then add water miscible organic solvent (EtOH (10mg/ml), MeOH, DMSO (10mg/ml), DMF) until solution is 50/50 base/solvent. When a clear solution is obtained, it can be diluted into an aqueous medium and titrated or buffered to any pH>7. Use immediately.

Formula: C₃₄H₃₄N₄O₄
MW: 562.66 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151109
CAS Number: 553-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dfdad1815fc213c5ae12bb42ac7a089942abf8d&vid=0d3ea22cfa0e6fc633e7c0b8ec3f181e948df087&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR