R59 949 ≥98%

Supplier: Enzo Life Sciences

430-028-M005 430-028-M025
89150-922EA 237 CAD
89150-922 89150-924
R59 949 ≥98%
R59 949

Potentiates PKC activity


Inhibitor of diacylglycerol kinase in intact cells (IC50=0.12µM) and in isolated membranes (IC50=0.3µM). More potent analog of R59 022 (Prod. No. ALX-430-027). Useful for elucidating roles of protein kinase C (PKC).

Formula: C₂₈H₂₅F₂N₃OS
MDL Number: MFCD00069258
CAS Number: 120166-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d5e20a40db69b5100d7abc89b4fac07742a04e6&vid=a399754fcba05014dac97f36ee80a0baa5b9af2e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR