Noc-12 ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Warning


Packaged under inert gas.

430-019-M005 430-019-M010
89150-902EA 156.42 CAD
89150-902 89150-904
Noc-12 ≥97% (by HPLC)
Noc-12

Nitric oxide (NO) donor.


  • Formulation: White powder.


The half-life time of 0.1 mM NOC-12 is 100 min at 37 °C in 0.1 M PBS, pH 7.4.


Soluble in 0.1 M NaOH (10 mg/mL).


Synonym: 1-Hydroxy-2-oxo-3-(N-ethyl-2-aminoethyl)-3-ethyl-1-triazene


Caution: Protect from light and moisture. 

Formula: C₆H₁₆N₄O₂
Storage Temperature: Freezer
CAS Number: 146724-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6533afad1dc7ce79c20a60ffbfc9c15a3b664dc&vid=3a313460e27dad47fa938e73d45689f9b2a7d37e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR