Noc-7 ≥96% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
430-018-M005
430-018-M010
89150-898EA
176.96
CAD
89150-898
89150-900
Noc-7 ≥96% (by HPLC)
Noc-7
Nitric oxide (NO) donor.
- Formulation: White to off-white crystalline solid.
The half-life time of 0.1 mM NOC-7 is 5 min at 37 °C in 0.1 M PBS, pH 7.4.
Reconstitute with 0.1 M sodium hydroxide solution to 10 - 50 mM.
Synonym: 1-Hydroxy-2-oxo-3-(N-3-methyl-aminopropyl)-3-methyl-1-triazene
Caution: Protect from light and moisture.
Formula:
C₅H₁₄N₄O₂ Storage Temperature: Freezer |
CAS Number:
146724-84-7 |
|
Specification Test Results
Purity | ≥96% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...