Noc-5 ≥90% (by HPLC)
Supplier: Enzo Life Sciences
|
Warning
|
Packaged under inert gas.
430-017-M005
430-017-M010
89150-894EA
268.6
CAD
89150-894
89150-896
Noc-5 ≥90% (by HPLC)
Noc-5
Nitric oxide (NO) donor.
- Formulation: White to off-white crystalline solid.
The half-life time of 0.1 mM NOC-5 is 25 min at 37 °C in 0.1 M PBS, pH 7.4.
Soluble in 0.1 M NaOH (10 mg/mL).
Synonym: 1-Hydroxy-2-oxo-3-(3-aminopropyl)-3-isopropyl-1-triazene
Caution: Protect from light and moisture.
Formula:
C₆H₁₆N₄O₂ Storage Temperature: Freezer |
CAS Number:
146724-82-5 |
|
Specification Test Results
Purity | ≥90% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...