PAPA NONOate ≥97%

Supplier: Enzo Life Sciences
Warning

430-016-5005 430-016-M005 430-016-M025
89150-888EA 360.24 CAD
89150-888 89150-890 89150-892
PAPA NONOate ≥97%
PAPA NONOate

Nitric oxide (NO) donor.


  • Formulation: White to off-white solid.


The half-life time of PAPA NONOate is 76.6 min at 22 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0.


Soluble in water or methanol. Relatively stable in alkaline solution.


Synonyms: PAPA/NO, NOC-15, (Z)-1-[N-(3-Ammoniopropyl)-N-(n-propyl)amino]diazen-1-ium-1,2-diolate


Caution: Protect from moisture. Keep under inert gas.

Formula: C₆H₁₆N₄O₂
MW: 176.2 g/mol
Storage Temperature: Freezer
CAS Number: 146672-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ad266d3af1daac5773fef90f66f1739b29c4a5a&vid=dc79f3e0d9b6322c9c59bd2cba9900e098654bd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97%
Identity Determined by IR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR