Purmorphamine ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine, 9-Cyclohexyl-N-(4-morpholinophenyl)-2-(naphthalen-1-yloxy)-9H-purin-6-amine

420-045-M001 420-045-M005 420-045-M025
89150-814EA 276.5 CAD
89150-814 89150-816 89150-818
Purmorphamine ≥98% (by HPLC)
Purmorphamine

Smoothened activator.


  • Formulation: Off-white to beige solid.


Cell permeable purine compound. Induces osteoblast differentiation. Agonist of smoothened by activation of the Hedgehog (Hh) pathway. Also shown to induce transdifferentiation in pre-adipocytes and myoblasts. Binds to and activates 7-transmembrane receptor of Hh.


Soluble in DMSO (10 mM).


Synonym: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine

Formula: C₃₁H₃₂N₆O₂
MW: 520.637 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09037557
CAS Number: 483367-10-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ede1a84d8efe630f46d249278f47238c3a9ae9&vid=db3a9b923ae691399ec81e13b8e81f649f7fb391&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR