Temozolomide ≥98%

Supplier: Enzo Life Sciences

Synonyms: 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide

420-044-M025 420-044-M100 420-044-M500
89150-808EA 184.86 CAD
89150-808 89150-810 89150-812
Temozolomide ≥98%
Temozolomide

AGT inhibitor


Antitumor and antiangiogenic compound. DNA alkylating agent. Induces G2/M arrest and apoptosis through adduction of a methyl group to O6 position of guanine in genomic DNA. Inactivates DNA repair protein O(6)-alkylguanine DNA alkyltransferase (AGT) in base excision repair (BER) pathway. Chemotherapeutic agent. Modifies drug resistance.

Formula: C₆H₆N₆O₂
MW: 194.15 g/mol
MDL Number: MFCD00866492
CAS Number: 85622-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b800dda60eae9a40fa853b1f84aa64251d8df36&vid=d9127fe573bae5a5e9498dbc62ec01ac12c52bb4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR