WS-12 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
AVX-012, (1R,2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexane-1-carboxamide
TRMP8 agonist.
- Formulation: White to off-white powder.
Cooling agent. High-affinity TRPM8 (cold menthol receptor 1; CMR1) agonist. Has an EC₅₀ value ~2000 times lower than that of (-)-menthol.
Soluble in 100% ethanol, methanol, ethyl acetate, dimethyl formamide or acetonitrile; insoluble in water or DMSO.
Synonym: (2S,5R)-2-Isopropyl-N-(4-methoxyphenyl)-5-methylcyclohexanecarboximide
Formula:
C₁₈H₂₇NO₂ MW: 289.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11100191 CAS Number: 68489-09-8 |
Specification Test Results
Purity | ≥98% (HPLC) |
Identity | Determined by ¹H-NMR and MS |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...