Gardiquimod ≥98% (by HPLC and UV method)

Supplier: Enzo Life Sciences
Danger

420-040-M025 420-040-M100
89150-790EA 334.96 CAD
89150-790 89150-792
Gardiquimod ≥98% (by HPLC and UV method)
Gardiquimod

TLR7 ligand.


  • Formulation: White to off-white solid.


Selective ligand for human or mouse toll-like receptor 7 (TLR7). Induces the activation of NF-κB in HEK 293 cells expressing TLR7. At high concentrations (3 µg/mL) slightly activates TLR8. More active than Imiquimod.


Soluble in DMSO (>40 mg/mL), dioxane (7 mg/mL), methanol (>40 mg/mL) or acetonitrile (2 mg/mL).

Formula: C₁₇H₂₃N₅O
Storage Temperature: Freezer
CAS Number: 1020412-43-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38667693c67565e9e010c026948a596f1ed63d70&vid=97f3dc2b807ea4252b7d76ec192e5ac251687ba2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC (UV))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR