Imiquimod

Supplier: Enzo Life Sciences

420-039-M100 420-039-M250
89150-786EA 142.2 CAD
89150-786 89150-788
Imiquimod
Imiquimod

TLR7 ligand


Topical immune response modifier that inhibits angiogenesis. Up-regulates IL-18 and down-regulates MMP-9 through recognition of Toll-like receptor 7 (TLR7) and subsequent activation of MyD88-dependent pathway.

Formula: C₁₄H₁₆N₄
MDL Number: MFCD00866946
CAS Number: 99011-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d4a8068781f42d1447ba38e453d781d261e60a&vid=2ff5c0a66cc3d0aaad73ce1a8a49cb632954bddc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR