Resiquimod (synthetic) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

420-038-M005 420-038-M025
89150-782EA 120.08 CAD
89150-782 89150-784
Resiquimod (synthetic) ≥98% (by HPLC)
Resiquimod

Ligand for TLR7 and TLR8.


  • Formulation: White to off-white solid.
  • Synthetic
  • Endotoxin-free


R-848 is a selective ligand for Toll-like receptor 7 (TLR7) in mouse and for TLR7 and TLR8 in human. Potent antitumor and antiviral compound. Stimulates antibody secretion, cytokine production, protection from apoptosis and CD80 upregulation.


Soluble in DMSO, dichloromethane, 100% or 5% methanol; poorly soluble in acetonitrile or ethyl acetate.

Formula: C₁₇H₂₂N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD00937759
CAS Number: 144875-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d512b5f40422decdfadc7c1fb5d4d2ccc0cbd44b&vid=9ce549863ff5b6090e064795fa646a9633522f93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by MS, ¹H-NMR and ¹³C-NMR
Source Synthetic
Endotoxin Content Endotoxin-free


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR