HAT activator, CTPB ≥97% (by 1H-NMR)

Supplier: Enzo Life Sciences

Synonyms: N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-ethoxy-6-pentadecylbenzamide, CTPB

420-033-M001 420-033-M005
89150-766EA 96.38 CAD
89150-766 89150-768
HAT activator, CTPB ≥97% (by 1H-NMR)
HAT activator, CTPB

p300 HAT activator.


  • Formulation: White to off-white crystalline solid.


Potent activator of p300 HAT (histone acetyltransferase), but not of PCAF (p300/CBP-associated factor) HAT activities.


Soluble in DMSO.


Synonym: N-(4-Chloro-3-trifluoromethyl-phenyl)-2-ethoxy-6-pentadecyl-benzamide

Formula: C₃₁H₄₃ClF₃NO₂
MW: 554.135 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08702707
CAS Number: 586976-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a140d7024d4799dc995037d5fa4a49fc79cee07&vid=14e0b8cea83026960c503135365b06e07b523df1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (¹H-NMR)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR