BAY 41-2272 ≥98%

Supplier: Enzo Life Sciences

420-030-M005 420-030-M025
89150-754EA 237 CAD
89150-754 89150-756
BAY 41-2272 ≥98%
Formula: C₂₀H₁₇FN₆
MDL Number: MFCD06411391
CAS Number: 256376-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b4aba70bafda9984eac260513422909fdffb726&vid=956b1fb70835b3192841ec4b2938932b6c732964&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR