YC-1 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

420-025-M001 420-025-M005
89150-734EA 71.1 CAD
89150-734 89150-736 89150-738
YC-1 ≥98% (by HPLC)
YC-1

Guanylyl cyclase activator


Nitric oxide (NO) independent, superoxide-sensitive stimulator of soluble guanylyl cyclase (sGC), EC50=18.6μM. Induces concentration-dependent increase in cGMP levels in cultured cells and in rabbit aortic rings. Inhibits platelet adhesion to collagen. Non-specific phosphodiesterase inhibitor. YC-1 inhibits HIF-alpha activity in tumors resulting in blocked angiogenesis and an inhibition of tumor growth. Inhibits Na+ channel.Binds to [3H]BW202W92.

Formula: C₁₉H₁₆N₂O₂
MDL Number: MFCD06407798
CAS Number: 170632-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e456bd0f6133423931e9de7e8b32d83ccfebf69&vid=16164239b9b2957bbf5f24430b8aff4043347d06&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR